Läs mer
Diethazine, LoGiCal
Produktkod. 30459191
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30459191 10 mg
1 options
Produktkod. 30459191 Leverantör LoGiCal Leverantörsnummer LGCFOR1593.01
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Diethazine
CAS 60-91-3
Analyt- eller komponentnamn Diethazine
Frakt skick Ice pack (-20°C)
Molekylformel C18 H22 N2 S
Certifikat och efterlevnad ISO 9001
Synonym 10H-Phenothiazine-10-ethanamine, N,N-diethyl-, Phenothiazine, 10-[2-(diethylamino)ethyl]- (8CI), 10-(2-Diethylaminoethyl)phenothiazine, Diethazine, Dolisina, Eazamine, Eazaminum, Ethylemin, N-(2'-Diethylaminoethyl)dibenzoparathiazine, N-(Diethylaminoethyl)thiodiphenylamine, Parkazin, RP 2987
InChI formel InChI=1S/C18H22N2S/c1-3-19(4-2)13-14-20-15-9-5-7-11-17(15)21-18-12-8-6-10-16(18)20/h5-12H,3-4,13-14H2,1-2H3
Lösningstyp Neat
LEDER CCN(CC)CCN1c2ccccc2Sc3ccccc13
IUPAC-namn N,N-diethyl-2-phenothiazin-10-ylethanamine
Industry Type Clinical Testing
Molekylvikt (g/mol) 298.45
Kvantitet 10 mg
Formel vikt 298.1504 g/mol