Learn More
Diethoxy(methyl)(phenyl)silane, 95%, BLD Pharm™
Specifications
Specifications
| CAS | 775-56-4 |
| Molecular Formula | C11H18O2Si |
| MDL Number | MFCD00048541 |
| Quantity | 100 g |
| SMILES | C[Si](OCC)(OCC)C1=CC=CC=C1 |
| Molecular Weight (g/mol) | 210.34 |
| Percent Purity | 95% |
| Chemical Name or Material | Diethoxy(methyl)(phenyl)silane |