missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl 1-propylphosphonate, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Description
Diethyl 1-propylphosphonate, is used as a medical intermediate and in organic synthesis. It is also used in electronics industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Diethyl 1-propylphosphonate, is used as a medical intermediate and in organic synthesis. It is also used in electronics industry.
Solubility
Soluble in water (27 g/L) (25°C).
Notes
Stable at room temperature in closed containers under normal storage and handling conditions. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to oxidizing agents, reducing agents, acids, bases.
Diethyl 1-propylphosphonate, is used as a medical intermediate and in organic synthesis. It is also used in electronics industry.
Solubility
Soluble in water (27 g/L) (25°C).
Notes
Stable at room temperature in closed containers under normal storage and handling conditions. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to oxidizing agents, reducing agents, acids, bases.
Specifications
Boiling Point | 94°C to 96°C (12 mmHg) |
Refractive Index | 1.419 |
Quantity | 10 g |
Beilstein | 1762076 |
Solubility Information | Soluble in water (27g/L) (25°C). |
Formula Weight | 180.19 |
Percent Purity | 97% |
Chemical Name or Material | Diethyl 1-propylphosphonate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'