Läs mer
Diethyl 2-Acetamido-2-[2-(4-octylphenylethyl)malonate, TRC
Produktkod. 30387073
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30387073 100 mg
1 options
Produktkod. 30387073 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD443125100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Diethyl 2-Acetamido-2-[2-(4-octylphenylethyl)malonate
CAS 162358-08-9
Molekylformel C25 H39 N O5
Kvantitet 100 mg
Synonym Propanedioic acid, 2-(acetylamino)-2-[2-(4-octylphenyl)ethyl]-, 1,3-diethyl ester, Propanedioic acid, (acetylamino)[2-(4-octylphenyl)ethyl]-, diethyl ester (9CI), (Acetylamino)[2-(4-octylphenyl)ethyl]propanedioic acid diethyl ester, 2-Acetylamino-2-(2-(4-octylphenyl)ethyl)propanedioic acid diethyl ester, Diethyl 2-(acetamido)-2-(2-(4-octylphenyl)ethyl)propanedioate, Diethyl 2-[2-(4-n-octylphenyl)ethyl]-2-acetamidomalonate
InChI formel InChI=1S/C25H39NO5/c1-5-8-9-10-11-12-13-21-14-16-22(17-15-21)18-19-25(26-20(4)27,23(28)30-6-2)24(29)31-7-3/h14-17H,5-13,18-19H2,1-4H3,(H,26,27)
LEDER CCCCCCCCc1ccc(CCC(NC(=O)C)(C(=O)OCC)C(=O)OCC)cc1
IUPAC-namn diethyl 2-acetamido-2-[2-(4-octylphenyl)ethyl]propanedioate
Molekylvikt (g/mol) 433.58
Formel vikt 433.2828