Läs mer
Diethyl 2-(Methylthio)-4,5-pyrimidinedicarboxylate, TRC
Produktkod. 30372385
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30372385 25 mg
1 options
Produktkod. 30372385 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD44457825MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Diethyl 2-(Methylthio)-4,5-pyrimidinedicarboxylate
CAS 149771-08-4
Molekylformel C11H14N2O4S
Kvantitet 25 mg
Synonym 2-Methylsulfanylpyrimidine-4,5-dicarboxylic Acid Diethyl Ester, NSC 165442
InChI formel InChI=1S/C11H14N2O4S/c1-4-16-9(14)7-6-12-11(18-3)13-8(7)10(15)17-5-2/h6H,4-5H2,1-3H3
LEDER CCOC(=O)c1cnc(SC)nc1C(=O)OCC
Molekylvikt (g/mol) 270.3
Formel vikt 270.0674