Läs mer
Diethyl (2S)-2-[[4-(Methylamino)benzoyl]amino]pentanedioate, Mikromol™
Produktkod. 17981616
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
17981616 25 mg
1 options
Produktkod. 17981616 Leverantör Mikromol Leverantörsnummer MM0040.180025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Diethyl (2S)-2-[[4-(Methylamino)benzoyl]amino]pentanedioate
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Diethyl (2S)-2-[(4-(Methylamino)benzoyl)amino]pentanedioate
CAS 2378-95-2
Analyt- eller komponentnamn Diethyl (2S)-2-[(4-(Methylamino)benzoyl)amino]pentanedioate
Frakt skick Room Temperature
Molekylformel C17 H24 N2 O5
Certifikat och efterlevnad ISO 9001
Synonym Diethyl (2S)-2-[[4-(Methylamino)benzoyl]amino]pentanedioate, L-Glutamic acid, N-[4-(methylamino)benzoyl]-, 1,5-diethyl ester, Glutamic acid, N-[p-(methylamino)benzoyl]-, diethyl ester (7CI), Glutamic acid, N-[p-(methylamino)benzoyl]-, diethyl ester, L- (8CI), L-Glutamic acid, N-[4-(methylamino)benzoyl]-, diethyl ester (9CI), Diethyl N-(p-N-methylaminobenzoyl)glutamate, Diethyl N-[4-(methylamino)benzoyl]-L-glutamate, N-(p-Methylaminobenzoyl)glutamic acid diethyl ester, Diethyl (2S)-2-[[4-(Methylamino)benzoyl]amino]pentanedioate
InChI formel InChI=1S/C17H24N2O5/c1-4-23-15(20)11-10-14(17(22)24-5-2)19-16(21)12-6-8-13(18-3)9-7-12/h6-9,14,18H,4-5,10-11H2,1-3H3,(H,19,21)/t14-/m0/s1
LEDER CCOC(=O)CC[C@H](NC(=O)c1ccc(NC)cc1)C(=O)OCC
IUPAC-namn diethyl (2S)-2-[[4-(methylamino)benzoyl]amino]pentanedioate
Molekylvikt (g/mol) 336.383
Kvantitet 25 mg
Formel vikt 336.169 g/mol