missing translation for 'onlineSavingsMsg'
Learn More

Diethyl (3,3-diethoxypropyl)phosphonate, 95%, stabilized with max. 1% w/w Na2CO3, Thermo Scientific Chemicals

Catalog Number p-3778251
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
15110-17-5
C11H25O5P
268.29
MFCD00800259
WKPXMGJOGYEGNW-UHFFFAOYSA-N
diethyl 3,3-diethoxypropyl phosphonate, diethyl 3,3-diethoxypropylphosphonate, phosphonic acid, p-3,3-diethoxypropyl-, diethyl ester, 3-diethoxyphosphoryl-1,1-diethoxy-propane, 3-diethoxyphosphoryl propanaldehyde diethyl acetal, phosphonic acid,p-3,3-diethoxypropyl-, diethyl ester
328856
3-diethoxyphosphoryl-1,1-diethoxypropane
CCOC(CCP(=O)(OCC)OCC)OCC
This item is not returnable. View return policy
15110-17-5
C11H25O5P
268.29
MFCD00800259
WKPXMGJOGYEGNW-UHFFFAOYSA-N
diethyl 3,3-diethoxypropyl phosphonate, diethyl 3,3-diethoxypropylphosphonate, phosphonic acid, p-3,3-diethoxypropyl-, diethyl ester, 3-diethoxyphosphoryl-1,1-diethoxy-propane, 3-diethoxyphosphoryl propanaldehyde diethyl acetal, phosphonic acid,p-3,3-diethoxypropyl-, diethyl ester
328856
3-diethoxyphosphoryl-1,1-diethoxypropane
CCOC(CCP(=O)(OCC)OCC)OCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY
Color Orange to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4290 to 1.4320
Quantity 5 g
Stabilizer 1% max. Na2CO3
Formula Weight 268.29
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Diethyl (3, 3-diethoxypropyl)phosphonate, Stabilized with max.1% (w/w) Na2CO3
Show More Show Less
compliance-icons
Product Identifier
  • Diethyl (3,3-diethoxypropyl)phosphonate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'