Läs mer
Diethyl 4,5-Thiazoledicarboxylate, TRC
Produktkod. 30378293
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30378293 50 mg
30378502 100 mg
30371276 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30378293 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD52294050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 18940-72-2
Molekylformel C9H11NO4S
Molekylvikt (g/mol) 229.25
Synonym 4,5-Thiazoledicarboxylic Acid Diethyl Ester (8CI,9CI)
IUPAC-namn diethyl 1,3-thiazole-4,5-dicarboxylate
LEDER CCOC(=O)c1ncsc1C(=O)OCC
Specifikationer
Kemiskt namn eller material Diethyl 4,5-Thiazoledicarboxylate
Kvantitet 50 mg
InChI formel InChI=1S/C9H11NO4S/c1-3-13-8(11)6-7(15-5-10-6)9(12)14-4-2/h5H,3-4H2,1-2H3
Formel vikt 229.0409