Läs mer
Diethyl a-Acetylglutarate, TRC
Produktkod. 30371456
Change view
Kvantitet:
25 g
Produktkod. Kvantitet
30371456 25 g
1 options
Produktkod. 30371456 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD44351025G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Diethyl alpha-Acetylglutarate
CAS 1501-06-0
Molekylformel C11H18O5
Kvantitet 25 g
Synonym 2-acetylglutaric Acid Diethyl ester, 2-acetylpentanedioic Acid Diethyl Ester, Diethyl 2-Acetylglutarate, Diethyl 2-Acetylpentanedioate, NSC 42535
InChI formel InChI=1S/C11H18O5/c1-4-15-10(13)7-6-9(8(3)12)11(14)16-5-2/h9H,4-7H2,1-3H3
LEDER CCOC(=O)CCC(C(=O)C)C(=O)OCC
IUPAC-namn diethyl 2-acetylpentanedioate
Molekylvikt (g/mol) 230.26
Formel vikt 230.1154