Learn More
Diethyl acetylmethylphosphonate, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Diethyl acetylmethylphosphonate is used as ligand in coordination chemistry to prepare lanthanide series metals like terbium, europium and neodymium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Diethyl acetylmethylphosphonate is used as ligand in coordination chemistry to prepare lanthanide series metals like terbium, europium and neodymium.
Solubility
Miscible with tetrahydrofuran, ether, dichloromethane and chloroform.
Notes
Incompatible with strong oxidizing agents.
Diethyl acetylmethylphosphonate is used as ligand in coordination chemistry to prepare lanthanide series metals like terbium, europium and neodymium.
Solubility
Miscible with tetrahydrofuran, ether, dichloromethane and chloroform.
Notes
Incompatible with strong oxidizing agents.
Specifications
Density | 1.01 |
Boiling Point | 129°C (9 mmHg) |
Flash Point | 113°C (235°F) |
Refractive Index | 1.433 |
Quantity | 1 g |
Solubility Information | Miscible with tetrahydrofuran,ether,dichloromethane and chloroform. |
Formula Weight | 194.17 |
Percent Purity | 97% |
Chemical Name or Material | Diethyl acetylmethylphosphonate |