Läs mer
Diethyl aminoethyl hexanoate citrate, MedChemExpress
Produktkod. 16405426
Encompass_Preferred
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
16405426 100 mg
1 options
Produktkod. 16405426 Leverantör Medchem Express Leverantörsnummer HY112106A
MedChemExpress Diethyl aminoethyl hexanoate citrate is a compound that is widely used as a plant growth regulator.
Diethyl aminoethyl hexanoate citrate is a compound that is widely used as a plant growth regulator.
Specifikationer
Kemiskt namn eller material Diethyl aminoethyl hexanoate citrate
Fysisk form Solid
CAS 220439-24-7
Färg White
Kvantitet 100 mg
Anteckningar om renhetsgrad Research
Molekylformel C18H33NO9
Synonym DA-6 citrate 2-Diethylaminoethyl hexanoate citrate
Löslighetsinformation DMSO : 100 mg/mL (245.42 mM; Need ultrasonic)
LEDER CCCCCC(OCCN(CC)CC)=O.OC(CC(C(O)=O)(O)CC(O)=O)=O
Molekylvikt (g/mol) 407.46
Formel vikt 407.46
Procent renhet 98.0%