missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl chlorophosphate, 97+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
A phosphorous halide used for biochemical research. Also used as an intermediate in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
A phosphorous halide used for biochemical research. Also used as an intermediate in organic synthesis.
Solubility
Soluble in alcohol and chloroform. Slightly soluble in water.
Notes
Moisture Sensitive. Store away from water/moisture. Store away from oxidizing agent.
A phosphorous halide used for biochemical research. Also used as an intermediate in organic synthesis.
Solubility
Soluble in alcohol and chloroform. Slightly soluble in water.
Notes
Moisture Sensitive. Store away from water/moisture. Store away from oxidizing agent.
Specifications
Density | 1.194 |
Boiling Point | 60°C (2 mmHg) |
Flash Point | 61°C (142°F) |
Quantity | 25 g |
UN Number | UN3278 |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in alcohol and chloroform. Slightly soluble in water. |
Formula Weight | 172.55 |
Percent Purity | ≥97% |
Chemical Name or Material | Diethyl chlorophosphate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'