Läs mer
Diethyl DL-Malate, TRC
Produktkod. 30007900
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30007900 5 g
1 options
Produktkod. 30007900 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4445855G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Diethyl DL-Malate
CAS 7554-12-3
Molekylformel C8 H14 O5
Kvantitet 5 g
Synonym 2-Hydroxybutanedioic Acid 1,4-Diethyl Ester, Hydroxybutanedioic Acid Diethyl Ester, Malic Acid Diethyl Ester, (±)-Diethyl Malate, Diethyl 2-Hydroxysuccinate, Diethyl Malate
InChI formel InChI=1S/C8H14O5/c1-3-12-7(10)5-6(9)8(11)13-4-2/h6,9H,3-5H2,1-2H3
LEDER CCOC(=O)CC(O)C(=O)OCC
IUPAC-namn diethyl 2-hydroxybutanedioate
Molekylvikt (g/mol) 190.19
Formel vikt 190.0841