Läs mer
Diethyl (E)-4-{2-[2-(tert-Butoxycarbonyl)vinyl]phenyl}-2,6-dimethylpyridine-3,5-dicarboxylate, Mikromol™
Produktkod. 30460054
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30460054 4 x 25 mg
30052263 25 mg
30051231 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30460054 Leverantör Mikromol Leverantörsnummer MM0407.0200254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Diethyl (E)-4-{2-[2-(tert-Butoxycarbonyl)vinyl]phenyl}-2,6-dimethylpyridine-3,5-dicarboxylate, Mikromol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Diethyl (E)-4-{2-[2-(tert-Butoxycarbonyl)vinyl]phenyl}-2,6-dimethylpyridine-3,5-dicarboxylate
CAS 130996-24-6
Analyt- eller komponentnamn Diethyl (E)-4-{2-[2-(tert-Butoxycarbonyl)vinyl]phenyl}-2,6-dimethylpyridine-3,5-dicarboxylate
Frakt skick Room Temperature
Molekylformel C26 H31 N O6
Certifikat och efterlevnad ISO 17025
Synonym Diethyl (E)-4-{2-[2-(tert-Butoxycarbonyl)vinyl]phenyl}-2,6-dimethylpyridine-3,5-dicarboxylate, Lacidipine Imp. B (BP), Lacidipine Impurity B
InChI formel InChI=1S/C26H31NO6/c1-8-31-24(29)21-16(3)27-17(4)22(25(30)32-9-2)23(21)19-13-11-10-12-18(19)14-15-20(28)33-26(5,6)7/h10-15H,8-9H2,1-7H3/b15-14+
Lösningstyp Neat
LEDER CCOC(=O)c1c(C)nc(C)c(C(=O)OCC)c1c2ccccc2\C=C\C(=O)OC(C)(C)C
IUPAC-namn diethyl 2,6-dimethyl-4-[2-[(E)-3-[(2-methylpropan-2-yl)oxy]-3-oxoprop-1-enyl]phenyl]pyridine-3,5-dicarboxylate
Molekylvikt (g/mol) 453.53
Kvantitet 4 x 25 mg
Formel vikt 453.2151 g/mol