missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl Ether, Anhydrous, Extra Dry, for Synthesis, Stabilised with Pyrogallol, Fisher Chemical™
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Packaging:
Amber Glass Bottle
Plastic-coated Glass Bottle
Description
Diethyl Ether, Anhydrous, Extra Dry, for Synthesis, Stabilised with Pyrogallol
Specifications
Melting Point | -116°C |
Color | Colorless |
Formula Weight | 74.12g/mol |
Boiling Point | 34.6°C |
Quantity | 500 mL |
Packaging | Amber Glass Bottle |
Physical Form | Liquid |
Vapor Pressure | 587mbar at 20°C |
Viscosity | 0.2448cp at 20°C |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'