Läs mer
Diethyl Ethylphenylmalonate, TRC
Produktkod. 30370950
Change view
Kvantitet:
10 g
Produktkod. Kvantitet
30370950 10 g
1 options
Produktkod. 30370950 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD44445510G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Diethyl Ethylphenylmalonate
CAS 76-67-5
Molekylformel C15H20O4
Kvantitet 10 g
Synonym Diethyl 2-ethyl-2-phenylmalonate, NSC 5631, Ethylphenylpropanedioic Acid Diethyl Ester
InChI formel InChI=1S/C15H20O4/c1-4-15(13(16)18-5-2,14(17)19-6-3)12-10-8-7-9-11-12/h7-11H,4-6H2,1-3H3
LEDER CCOC(=O)C(CC)(C(=O)OCC)c1ccccc1
Molekylvikt (g/mol) 264.32
Formel vikt 264.1362