missing translation for 'onlineSavingsMsg'
Learn More

Diethyl formamidomalonate, 97%, ACROS Organics™

Brand:  Acros Organics 114370050

Additional Details : CAS Number : 6326-44-9 Weight : 0.00500kg

Packaging Glass bottle
Quantity 5g

 View more versions of this product

Product Code. 15219616

  • 281.00 SEK / 5g

Estimated Shipment: 05-07-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

6326-44-9
203.19
PFLHGSJLYNJIOF-UHFFFAOYSA-N
80617
CCOC(=O)C(C(=O)OCC)NC=O
C8H13NO5
MFCD00003279
diethyl formamidomalonate, diethyl 2-formamidomalonate, diethyl 2-formylaminomalonate, diethyl formaminomalonate, diethyl2-formylaminomalonate, 1,3-diethyl 2-formamidopropanedioate, diethyl 2-carbonylaminopropane-1,3-dioate, formamidomalonic acid, diethyl ester, malonic acid, formamido-, diethyl ester, diethylformaminomalonate
diethyl 2-formamidopropanedioate

Specifications

Diethyl formamidomalonate
Authentic
173.0°C to 176.0°C (11.0mmHg)
51.0°C to 56.0°C
97%
96% min. (GC)
C2H5O2CCH(NHCHO)CO2C2H5
04,47
PFLHGSJLYNJIOF-UHFFFAOYSA-N
diethyl 2-formamidopropanedioate
80617
Adhering Crystalline Powder
113°C
Glass bottle
White
5g
6326-44-9
C8H13NO5
MFCD00003279
diethyl formamidomalonate, diethyl 2-formamidomalonate, diethyl 2-formylaminomalonate, diethyl formaminomalonate, diethyl2-formylaminomalonate, 1,3-diethyl 2-formamidopropanedioate, diethyl 2-carbonylaminopropane-1,3-dioate, formamidomalonic acid, diethyl ester, malonic acid, formamido-, diethyl ester, diethylformaminomalonate
CCOC(=O)C(C(=O)OCC)NC=O
203.19
203.19
97%
Videos
SDS
Documents
Certificates