Learn More
Diethyl formamidomalonate, 97%, Thermo Scientific™
Catalog Number p-108988
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
6326-44-9
C8H13NO5
203.19
MFCD00003279
PFLHGSJLYNJIOF-UHFFFAOYSA-N
80617
diethyl 2-formamidopropanedioate
CCOC(=O)C(C(=O)OCC)NC=O
6326-44-9
C8H13NO5
203.19
MFCD00003279
PFLHGSJLYNJIOF-UHFFFAOYSA-N
80617
diethyl 2-formamidopropanedioate
CCOC(=O)C(C(=O)OCC)NC=O
Melting Point 51.0°C to 56.0°C
Color White
Boiling Point 173.0°C to 176.0°C (11.0 mmHg)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C2H5O2CCH(NHCHO)CO2C2H5
Quantity 5 g
Beilstein 04,47
Synonym diethyl formamidomalonate, diethyl 2-formamidomalonate, diethyl 2-formylaminomalonate, diethyl formaminomalonate, diethyl2-formylaminomalonate, 1,3-diethyl 2-formamidopropanedioate, diethyl 2-carbonylaminopropane-1,3-dioate, formamidomalonic acid, diethyl ester, malonic acid, formamido-, diethyl ester, diethylformaminomalonate
Formula Weight 203.19
Percent Purity 97%
Physical Form Adhering Crystalline Powder
Chemical Name or Material Diethyl formamidomalonate, 97%