missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl hydroxymethylphosphonate, tech. 85%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
Diethyl hydroxymethylphosphonate acts as a reactant for synthesis of α And β-dialkoxyphosphoryl isothiocyanate used in antiproliferative studies, dendritic polyglycerol anions for L-selectin inhibition, phosphonates for anti-HIV-1 activity, homo and copolymers of phosphonated norbornenes, Mitsunobu reactions. It is involved in studies of FBPase inhibition by phosphonic acid-containing thaizoles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Diethyl hydroxymethylphosphonate acts as a reactant for synthesis of α And β-dialkoxyphosphoryl isothiocyanate used in antiproliferative studies, dendritic polyglycerol anions for L-selectin inhibition, phosphonates for anti-HIV-1 activity, homo and copolymers of phosphonated norbornenes, Mitsunobu reactions. It is involved in studies of FBPase inhibition by phosphonic acid-containing thaizoles.
Notes
Incompatible with oxidizing agents. Store in a cool, dry condition in a well sealed container.
Diethyl hydroxymethylphosphonate acts as a reactant for synthesis of α And β-dialkoxyphosphoryl isothiocyanate used in antiproliferative studies, dendritic polyglycerol anions for L-selectin inhibition, phosphonates for anti-HIV-1 activity, homo and copolymers of phosphonated norbornenes, Mitsunobu reactions. It is involved in studies of FBPase inhibition by phosphonic acid-containing thaizoles.
Notes
Incompatible with oxidizing agents. Store in a cool, dry condition in a well sealed container.
Specifications
Density | 1.14 |
Boiling Point | 124°C to 126°C (3 mmHg) |
Flash Point | 110°C (230°F) |
Refractive Index | 1.444 |
Quantity | 5 g |
Formula Weight | 168.13 |
Percent Purity | 85% |
Grade | Technical |
Chemical Name or Material | Diethyl hydroxymethylphosphonate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'