Läs mer
Diethyl Malonate-13C3, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Diethyl Malonate-13C3 |
| CAS | 53051-81-3 |
| Molekylformel | C413C3H12O4 |
| Kvantitet | 25 mg |
| Synonym | Propanedioic Acid 1,3-Diethyl Ester-13C3, Malonic Acid Diethyl Ester-13C3, Propanedioic Acid Diethyl Ester-13C3, Carbethoxyacetic Ester-13C3, Dicarbethoxymethane-13C3, Diethyl Propane-1,3-dioate-13C3, Diethyl Propanedioate-13C3, Ethyl Malonate-13C3, Malonate Diethyl Ester-13C3, Methanedicarboxylic Acid Diethyl Ester-13C3, NSC 136903-13C3, NSC 8864-13C3 |
| InChI formel | InChI=1S/C7H12O4/c1-3-10-6(8)5-7(9)11-4-2/h3-5H2,1-2H3/i5+1,6+1,7+1 |
| LEDER | O=[13C]([13C]([H])([H])[13C](OCC)=O)OCC |
| Molekylvikt (g/mol) | 163.15 |