Learn More
Diethyl Malonate, TRC
Product Code. 30006898
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
Product Code. Quantity Unit Size
30006898 10 g 10g
1 options
Product Code. 30006898 Supplier Toronto Research Chemicals Supplier No. TRCD44442010G
 Restricted Item
Beräknad leverans: 03-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
CAS 105-53-3
Molecular Formula C7H12O4
Quantity 10 g
Synonym Propanedioic Acid 1,3-Diethyl Ester, Malonic Acid Diethyl Ester, Propanedioic Acid Diethyl Ester, Carbethoxyacetic Ester, Dicarbethoxymethane, Diethyl Propane-1,3-dioate, Diethyl Propanedioate, Ethyl Malonate, Malonate Diethyl Ester, Methanedicarboxylic Acid Diethyl Ester-, NSC 136903, NSC 8864;
InChI Formula InChI=1S/C7H12O4/c1-3-10-6(8)5-7(9)11-4-2/h3-5H2,1-2H3
SMILES CCOC(=O)CC(=O)OCC
Molecular Weight (g/mol) 160.17
Formula Weight 160.0736
Percent Purity >95%
Chemical Name or Material Diethyl Malonate
Grade GC