Läs mer
Diethyl Methylphenylmalonate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Diethyl Methylphenylmalonate |
| CAS | 34009-61-5 |
| Molekylformel | C4 H4 Cl N S |
| Kvantitet | 500 mg |
| Synonym | 2-Methyl-2-phenyl-propanedioic Acid 1,3-Diethyl Ester, Methylphenyl-malonic Acid Diethyl Ester, Methylphenyl-propanedioic Acid Diethyl Ester, Diethyl 2-Methyl-2-phenylmalonate, NSC 147481 |
| InChI formel | InChI=1S/C4H4ClNS/c5-3-4-6-1-2-7-4/h1-2H,3H2 |
| LEDER | ClCc1nccs1 |
| IUPAC-namn | 2-(chloromethyl)-1,3-thiazole |
| Molekylvikt (g/mol) | 133.599 |
| Formel vikt | 132.975 |