Läs mer
Diethyl Methylphosphonate-13C, TRC
Produktkod. 30376505
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30376505 5 mg
1 options
Produktkod. 30376505 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4423575MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Diethyl Methylphosphonate-13C
CAS 1342310-32-0
Molekylformel C413CH13O3P
Kvantitet 5 mg
Synonym Bis(diethoxyphosphinyl)methane-13C, DEMP-13C, Diethoxymethylphosphine Oxide-13C, Diethyl Methanephosphonate-13C, O,O-Diethyl Methylphosphonate-13C, P-Methyl-phosphonic Acid Diethyl Ester-13C
InChI formel InChI=1S/C5H13O3P/c1-4-7-9(3,6)8-5-2/h4-5H2,1-3H3/i3+1
LEDER [13CH3]P(=O)(OCC)OCC
Molekylvikt (g/mol) 153.12