Läs mer
Diethyl Oxalopropionate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Diethyl Oxalopropionate |
| CAS | 759-65-9 |
| Molekylformel | C9H14O5 |
| Kvantitet | 5 g |
| Synonym | 2-Methyl-3-oxo-butanedioic Acid 1,4-Diethyl Ester, Methyloxo-butanedioic Acid Diethyl Ester, Methyl-oxalacetic Acid Diethyl Ester, 2-Methyl-3-(oxo)succinic Acid Diethyl Ester, Diethyl 2-Methyl-3-oxo-1,4-butanedioate, Diethyl 2-Methyl-3-oxobutanedioate, Diethyl 2-Methyl-3-oxosuccinate, Diethyl 2-Oxo-3-methylsuccinate, Diethyl 3-Methyl-2-oxosuccinate, Diethyl Methyloxalacetate, Ethyl alpha-Ethoxalylpropionate, NSC 33946 |
| InChI formel | InChI=1S/C9H14O5/c1-4-13-8(11)6(3)7(10)9(12)14-5-2/h6H,4-5H2,1-3H3 |
| LEDER | CCOC(=O)C(C)C(=O)C(=O)OCC |
| IUPAC-namn | diethyl 2-methyl-3-oxobutanedioate |
| Molekylvikt (g/mol) | 202.2 |
| Formel vikt | 202.0841 |