Läs mer
Diethyl Phosphate-13C4 Sodium Salt, TRC
Produktkod. 30138081
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138081 10 mg
17994765 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30138081 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD44472210MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1329613-90-2
Molekylformel 13C4H10NaO4P
Molekylvikt (g/mol) 180.05
Synonym Phosphoric Acid Diethyl Ester-13C4 Sodium Salt, Ethyl Sodium Phosphate-13C4, Diethyl-13C4 Sodium Phosphate, Sodium Diethyl Phosphate-13C4;
LEDER O=P(O[13CH2][13CH3])(O[Na])O[13CH2][13CH3]
Specifikationer
Kemiskt namn eller material Diethyl Phosphate-13C4 Sodium Salt
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C4H11O4P.Na/c1-3-7-9(5,6)8-4-2;/h3-4H2,1-2H3,(H,5,6);/q;+1/p-1/i1+1,2+1,3+1,4+1;
Formel vikt 180.0349
Procent renhet >95%