Läs mer
Diethyl Suberate, TRC
Produktkod. 30371100
Change view
Kvantitet:
100 g
Produktkod. Kvantitet
30371100 100 g
1 options
Produktkod. 30371100 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD445000100G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Diethyl Suberate
CAS 2050-23-9
Molekylformel C12H22O4
Kvantitet 100 g
Synonym Octanedioic Acid 1,8-Diethyl Ester, Octanedioic Acid Diethyl Ester, Suberic Acid Diethyl Ester, Diethyl Octanedioate, NSC 62701
InChI formel InChI=1S/C12H22O4/c1-3-15-11(13)9-7-5-6-8-10-12(14)16-4-2/h3-10H2,1-2H3
LEDER CCOC(=O)CCCCCCC(=O)OCC
IUPAC-namn diethyl octanedioate
Molekylvikt (g/mol) 230.3
Formel vikt 230.1518