Läs mer
Diethyl Succinate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Succinic acid diethyl ester |
| CAS | 123-25-1 |
| Molekylformel | C8 H14 O4 |
| Kvantitet | 25 g |
| Synonym | Succinic acid-diethyl ester, Butanedioic acid, 1,4-diethyl ester (ACI), Butanedioic acid, diethyl ester (9CI), Succinic acid, diethyl ester (6CI, 8CI), 1,4-Diethyl butanedioate, Butandioic acid diethyl ester, Clorius, DESU, Diethyl 1,2-ethanedicarboxylate, Diethyl butanedioate, Diethyl succinate, Ethyl succinate, NSC 8875 |
| InChI formel | InChI=1S/C8H14O4/c1-3-11-7(9)5-6-8(10)12-4-2/h3-6H2,1-2H3 |
| LEDER | CCOC(=O)CCC(=O)OCC |
| IUPAC-namn | diethyl butanedioate |
| Molekylvikt (g/mol) | 174.19 |
| Formel vikt | 174.0892 |