missing translation for 'onlineSavingsMsg'
Learn More

Diethyl sulfate, 99%, Thermo Scientific Chemicals

Catalog Number p-3735844
Click to view available options
Quantity:
25 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
64-67-5
C4H10O4S
154.18
MFCD00009099
DENRZWYUOJLTMF-UHFFFAOYSA-N
sulfuric acid, diethyl ester, diethyl sulphate, diethylsulfate, diaethylsulfat, sulfuric acid diethyl ester, diethyl tetraoxosulfate, diaethylsulfat german, unii-k0fo4vfa7i, des van, diethylester kyseliny sirove
6163
CHEBI:34699
diethyl sulfate
CCOS(=O)(=O)OCC
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
64-67-5
C4H10O4S
154.18
MFCD00009099
DENRZWYUOJLTMF-UHFFFAOYSA-N
sulfuric acid, diethyl ester, diethyl sulphate, diethylsulfate, diaethylsulfat, sulfuric acid diethyl ester, diethyl tetraoxosulfate, diaethylsulfat german, unii-k0fo4vfa7i, des van, diethylester kyseliny sirove
6163
CHEBI:34699
diethyl sulfate
CCOS(=O)(=O)OCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
TRUSTED_SUSTAINABILITY
Melting Point -24.0°C
Density 1.1795g/mL
Boiling Point 208.0°C
Flash Point 104°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (C2H5O)2SO2
Refractive Index 1.3990 to 1.4010
Quantity 25 mL
Beilstein 01,327
Fieser 01,253
Merck Index 15,3149
Specific Gravity 1.1795
Solubility Information Solubility in water: 5g/L (20°C). Other solubilities: decomposes in water,soluble in alcohol and ether
Viscosity 1.79 mPa.s (20°C)
Formula Weight 154.18
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Diethyl sulfate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'