Läs mer
Diethyl Sulfone, TRC
Produktkod. 17958594
Change view
Kvantitet:
2.5 g
25 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17958594 2.5 g
17930095 25 g
17909454 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17958594 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4450102.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 597-35-3
Molekylformel C4 H10 O2 S
Molekylvikt (g/mol) 122.19
Synonym Ethane, 1,1'-sulfonylbis-, Ethyl sulfone (6CI,7CI,8CI), Diethyl sulfone, NSC 145719
IUPAC-namn 1-ethylsulfonylethane
LEDER CCS(=O)(=O)CC
Specifikationer
Kemiskt namn eller material Diethyl Sulfone
Kvantitet 2.5 g
InChI formel InChI=1S/C4H10O2S/c1-3-7(5,6)4-2/h3-4H2,1-2H3
Formel vikt 122.0402