missing translation for 'onlineSavingsMsg'
Learn More

Diethyl tert-butylmalonate, 97%, Thermo Scientific Chemicals

Catalog Number 11337246
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
759-24-0
C11H20O4
216.28
MFCD00009152
RJNICNBRGVKNSR-UHFFFAOYSA-N
diethyl tert-butylmalonate, diethyl t-butylmalonate, diethyl 1,1-dimethylethyl malonate, diethyl 2-tert-butyl malonate, propanedioic acid, 1,1-dimethylethyl-, diethyl ester, diethyltert-butylmalonate, diethyl 2-tert-butylmalonate #, tert-butylmalonic acid diethyl ester, 1,3-diethyl 2-tert-butylpropanedioate
69798
diethyl 2-tert-butylpropanedioate
CCOC(=O)C(C(=O)OCC)C(C)(C)C
This item is not returnable. View return policy
759-24-0
C11H20O4
216.28
MFCD00009152
RJNICNBRGVKNSR-UHFFFAOYSA-N
diethyl tert-butylmalonate, diethyl t-butylmalonate, diethyl 1,1-dimethylethyl malonate, diethyl 2-tert-butyl malonate, propanedioic acid, 1,1-dimethylethyl-, diethyl ester, diethyltert-butylmalonate, diethyl 2-tert-butylmalonate #, tert-butylmalonic acid diethyl ester, 1,3-diethyl 2-tert-butylpropanedioate
69798
diethyl 2-tert-butylpropanedioate
CCOC(=O)C(C(=O)OCC)C(C)(C)C

Diethyl tert-butylmalonate is used in the preparation of enantiomers of 4-tert-butyl-3-isopropyl-2,6,7-trioxa-1-phosphabicyclo[2.2.2]octane 1-sulfide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl tert-butylmalonate is used in the preparation of enantiomers of 4-tert-butyl-3-isopropyl-2,6,7-trioxa-1-phosphabicyclo[2.2.2]octane 1-sulfide.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY
Density 0.978
Boiling Point 102°C to 104°C (11 mmHg)
Flash Point 90°C (194°F)
Refractive Index 1.425
Quantity 1 g
Beilstein 1781709
Solubility Information Immiscible with water.
Formula Weight 216.28
Percent Purity 97%
Chemical Name or Material Diethyl tert-butylmalonate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'