Learn More
Diethyl tert-butylmalonate, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Diethyl tert-butylmalonate is used in the preparation of enantiomers of 4-tert-butyl-3-isopropyl-2,6,7-trioxa-1-phosphabicyclo[2.2.2]octane 1-sulfide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Diethyl tert-butylmalonate is used in the preparation of enantiomers of 4-tert-butyl-3-isopropyl-2,6,7-trioxa-1-phosphabicyclo[2.2.2]octane 1-sulfide.
Solubility
Immiscible with water.
Notes
Incompatible with strong oxidizing agents and strong bases.
Diethyl tert-butylmalonate is used in the preparation of enantiomers of 4-tert-butyl-3-isopropyl-2,6,7-trioxa-1-phosphabicyclo[2.2.2]octane 1-sulfide.
Solubility
Immiscible with water.
Notes
Incompatible with strong oxidizing agents and strong bases.
Specifications
Density | 0.978 |
Boiling Point | 102°C to 104°C (11 mmHg) |
Flash Point | 90°C (194°F) |
Refractive Index | 1.425 |
Quantity | 1 g |
Beilstein | 1781709 |
Solubility Information | Immiscible with water. |
Formula Weight | 216.28 |
Percent Purity | 97% |
Chemical Name or Material | Diethyl tert-butylmalonate |