missing translation for 'onlineSavingsMsg'
Learn More

Diethyl tetrafluorosuccinate, 97%, Thermo Scientific Chemicals

Catalog Number p-7046820
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
377-71-9
C8H10F4O4
246.158
MFCD00015155
BLZSSBPZHBFNBN-UHFFFAOYSA-N
diethyl tetrafluorosuccinate, diethyl perfluorosuccinate, butanedioic acid, tetrafluoro-, diethyl ester, diethyl 2,2,3,3-tetrafluorosuccinate, butanedioic acid, 2,2,3,3-tetrafluoro-, 1,4-diethyl ester, 1,4-diethyl 2,2,3,3-tetrafluorobutanedioate, diethyl 2,2,3,3-tetrafluorobutane-1,4-dioate, diethyltetrafluorosuccinate, ethyl perfluorosuccinate, acmc-1cqkj
67834
diethyl 2,2,3,3-tetrafluorobutanedioate
CCOC(=O)C(C(C(=O)OCC)(F)F)(F)F
This item is not returnable. View return policy
377-71-9
C8H10F4O4
246.158
MFCD00015155
BLZSSBPZHBFNBN-UHFFFAOYSA-N
diethyl tetrafluorosuccinate, diethyl perfluorosuccinate, butanedioic acid, tetrafluoro-, diethyl ester, diethyl 2,2,3,3-tetrafluorosuccinate, butanedioic acid, 2,2,3,3-tetrafluoro-, 1,4-diethyl ester, 1,4-diethyl 2,2,3,3-tetrafluorobutanedioate, diethyl 2,2,3,3-tetrafluorobutane-1,4-dioate, diethyltetrafluorosuccinate, ethyl perfluorosuccinate, acmc-1cqkj
67834
diethyl 2,2,3,3-tetrafluorobutanedioate
CCOC(=O)C(C(C(=O)OCC)(F)F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.273
Boiling Point 78°C (5 mmHg)
Refractive Index 1.368
Quantity 1 g
Beilstein 1798175
Formula Weight 246.16
Percent Purity 97%
Chemical Name or Material Diethyl tetrafluorosuccinate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'