Läs mer
Diethylcarbamazine N-Oxide, TRC
Produktkod. 30376464
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30376464 250 mg
1 options
Produktkod. 30376464 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD443910250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Diethylcarbamazine N-Oxide
CAS 34812-73-2
Molekylformel C10 H21 N3 O2
Kvantitet 250 mg
Synonym 1-Piperazinecarboxamide, N,N-diethyl-4-methyl-, 4-oxide, 1-Diethylcarbamyl-4-methylpiperazine 4-oxide, Diethylcarbamazine N-oxide, Diethylcarbamazine N-Oxide
InChI formel InChI=1S/C10H21N3O2/c1-4-11(5-2)10(14)12-6-8-13(3,15)9-7-12/h4-9H2,1-3H3
LEDER CCN(CC)C(=O)N1CC[N+](C)([O-])CC1
IUPAC-namn N,N-diethyl-4-methyl-4-oxidopiperazin-4-ium-1-carboxamide
Molekylvikt (g/mol) 215.29
Formel vikt 215.1634