missing translation for 'onlineSavingsMsg'
Learn More

Diethyl (hydroxymethyl)phosphonate, 97%, Thermo Scientific Chemicals

Catalog Number 10540214
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
3084-40-0
C5H13O4P
168.13
MFCD00014418
RWIGWWBLTJLKMK-UHFFFAOYSA-N
diethyl hydroxymethyl phosphonate, diethyl hydroxymethylphosphonate, phosphonic acid, hydroxymethyl-, diethyl ester, hydroxymethyl phosphonic acid, diethyl ester, hydroxymethyl phosphonic acid diethyl ester, phosphonic acid, p-hydroxymethyl-, diethyl ester, acmc-1corl, diethoxyphosphoryl-methanol
76513
diethoxyphosphorylmethanol
CCOP(=O)(CO)OCC
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
3084-40-0
C5H13O4P
168.13
MFCD00014418
RWIGWWBLTJLKMK-UHFFFAOYSA-N
diethyl hydroxymethyl phosphonate, diethyl hydroxymethylphosphonate, phosphonic acid, hydroxymethyl-, diethyl ester, hydroxymethyl phosphonic acid, diethyl ester, hydroxymethyl phosphonic acid diethyl ester, phosphonic acid, p-hydroxymethyl-, diethyl ester, acmc-1corl, diethoxyphosphoryl-methanol
76513
diethoxyphosphorylmethanol
CCOP(=O)(CO)OCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 1.1400g/mL
Boiling Point 124.0°C to 126.0°C (3.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula HOCH2P(O)(OC2H5)2
Refractive Index 1.4370 to 1.4410
Quantity 25 g
Specific Gravity 1.14
Formula Weight 168.13
Percent Purity 97%
Physical Form Viscous Liquid
Chemical Name or Material Diethyl (hydroxymethyl)phosphonate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'