Läs mer
Diethylene Glycol Cyclic Phthalate, TRC
Produktkod. 30136700
Change view
Kvantitet:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136700 10 g
17997144 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30136700 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD44431010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 13988-26-6
Molekylformel C12 H12 O5
Molekylvikt (g/mol) 236.22
Synonym 2,5,8-Benzotrioxacycloundecin-1,9-dione, 3,4,6,7-tetrahydro- (9CI, ACI), 3,4,6,7-Tetrahydro-2,5,8-benzotrioxacycloundecin-1,9-dione (ACI), Diethylene glycol, cyclic phthalate (8CI), Phthalic acid, cyclic oxydiethylene ester (8CI), 3,4,6,7-Tetrahydrobenzo[i][1,4,7]trioxacycloundecine-1,9-dione, 3,6,9-Trioxabicyclo[9.4.0]pentadeca-1(15),11,13-triene-2,10-dione, Diethylene glycol bisphthalate, Phthalic Acid, Cyclic Oxydiethylene Ester
IUPAC-namn 3,6,9-trioxabicyclo[9.4.0]pentadeca-1(11),12,14-triene-2,10-dione
LEDER O=C1OCCOCCOC(=O)c2ccccc12
Specifikationer
Kemiskt namn eller material Diethylene Glycol Cyclic Phthalate
Anteckningar om renhetsgrad HPLC
Kvantitet 10 g
InChI formel InChI=1S/C12H12O5/c13-11-9-3-1-2-4-10(9)12(14)17-8-6-15-5-7-16-11/h1-4H,5-8H2
Formel vikt 236.0685
Procent renhet >95%