missing translation for 'onlineSavingsMsg'
Learn More

Diethylene glycol monoethyl ether, 98%, Thermo Scientific Chemicals

Catalog Number p-7022068
Click to view available options
Quantity:
10,000 g
2500 g
500 g
This item is not returnable. View return policy
111-90-0
C6H14O3
134.175
MFCD00002872
XXJWXESWEXIICW-UHFFFAOYSA-N
diethylene glycol monoethyl ether, 2-2-ethoxyethoxy ethanol, carbitol, transcutol, ethoxy diglycol, dioxitol, ethyl carbitol, ethyl digol, carbitol solvent, ethanol, 2-2-ethoxyethoxy
8146
CHEBI:40572
2-(2-ethoxyethoxy)ethanol
CCOCCOCCO
This item is not returnable. View return policy
111-90-0
C6H14O3
134.175
MFCD00002872
XXJWXESWEXIICW-UHFFFAOYSA-N
diethylene glycol monoethyl ether, 2-2-ethoxyethoxy ethanol, carbitol, transcutol, ethoxy diglycol, dioxitol, ethyl carbitol, ethyl digol, carbitol solvent, ethanol, 2-2-ethoxyethoxy
8146
CHEBI:40572
2-(2-ethoxyethoxy)ethanol
CCOCCOCCO

Diethylene glycol monoethyl ether has low setting point and low viscocity at low temperature so it is used in manufacturing brake fluid. It is used as a flow and gloss promoter in paint industries, in production of printing ink and as a cleaner in offset printing. Also used in textile as a solvent for dyestuff in the printing and dying of fiber & fabrics, in the production & wood preservaties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethylene glycol monoethyl ether has low setting point and low viscocity at low temperature so it is used in manufacturing brake fluid. It is used as a flow and gloss promoter in paint industries, in production of printing ink and as a cleaner in offset printing. Also used in textile as a solvent for dyestuff in the printing and dying of fiber & fabrics, in the production & wood preservaties.

Solubility
Fully miscible in water.

Notes
It is hygroscopic. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point -80°C
Density 0.989
Boiling Point 201°C to 202°C
Flash Point 96°C (204°F)
Odor Pleasant
Linear Formula HOCH2CH2OCH2CH2OCH2CH3
Refractive Index 1.428
Quantity 500 g
Beilstein 1736441
Sensitivity Hygroscopic
Merck Index 14,1800
Solubility Information Fully miscible in water.
Formula Weight 134.18
Percent Purity 98%
Chemical Name or Material Diethylene glycol monoethyl ether
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'