missing translation for 'onlineSavingsMsg'
Läs mer

Diethylenetriamine, Sigma-Aldrich™

Produktkod. 15898980
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 mL
1 L
Förpackningsstorlek:
1 L
100ml
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet Förpackningsstorlek
15898980 100 mL 100ml
15866360 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 15898980 Leverantör Sigma Aldrich Leverantörsnummer 8032740100
 Begränsad artikel
Beräknad leverans: 26-11-2026
Lägg i varukorgen
Lägg i varukorgen

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

For synthesis

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 111-40-0

Specifikationer

Smältpunkt -35°C
Densitet 0.95 g/cm3 at 20°C
Kokpunkt 206°C to 209°C/1013 hPa
Molekylformel (H2NCH2CH2)2NH
MDL-nummer MFCD00008171
Kvantitet 100 mL
Synonym Diethylenetriamine, 2,2′-Iminobis (ethylamine), Bis(2-aminoethyl)amine, 2,2′-Diaminodiethylamine, Diethylenetriamine
Molekylvikt (g/mol) 103.17
Kvalitet Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.