Learn More
Diethyl succinate, 98%, Thermo Scientific Chemicals
Catalog Number p-3735846
530.17 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 kg
5 kg
25 kg
Packaging:
Glass Bottle
Plastic Can
Plastic drum
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
123-25-1
C8H14O4
174.2
MFCD00009208
DKMROQRQHGEIOW-UHFFFAOYSA-N
diethyl succinate, ethyl succinate, butanedioic acid, diethyl ester, 1,4-diethyl butanedioate, succinic acid, diethyl ester, succinic acid diethyl ester, diethyl ethanedicarboxylate, unii-elp55c13dr, diethylester kyseliny jantarove, fema no. 2377
31249
diethyl butanedioate
CCOC(=O)CCC(=O)OCC
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
123-25-1
C8H14O4
174.2
MFCD00009208
DKMROQRQHGEIOW-UHFFFAOYSA-N
diethyl succinate, ethyl succinate, butanedioic acid, diethyl ester, 1,4-diethyl butanedioate, succinic acid, diethyl ester, succinic acid diethyl ester, diethyl ethanedicarboxylate, unii-elp55c13dr, diethylester kyseliny jantarove, fema no. 2377
31249
diethyl butanedioate
CCOC(=O)CCC(=O)OCC
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Acidity 0.2mg KOH/g max.
Melting Point -20.0°C
Color Colorless to Yellow
Density 1.0400g/mL
Boiling Point 217.0°C
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Linear Formula C2H5O2CCH2CH2CO2C2H5
Refractive Index 1.4190 to 1.4210
Quantity 1 kg
Beilstein 02,609
Merck Index 15,8999
Specific Gravity 1.04
Solubility Information Solubility in water: slightly soluble in water. Other solubilities: miscible with alcohol and ether
Formula Weight 174.2
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Diethyl succinate