Läs mer
Diethylisobutyroylmalonate, TRC
Produktkod. 30378445
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30378445 500 mg
30370193 50 mg
30378988 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30378445 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD496335500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 21633-78-3
Molekylformel C11H18O5
Molekylvikt (g/mol) 230.25
IUPAC-namn diethyl 2-(2-methylpropanoyl)propanedioate
LEDER CCOC(=O)C(C(=O)OCC)C(=O)C(C)C
Specifikationer
Kemiskt namn eller material Diethylisobutyroylmalonate
Kvantitet 500 mg
InChI formel InChI=1S/C11H18O5/c1-5-15-10(13)8(9(12)7(3)4)11(14)16-6-2/h7-8H,5-6H2,1-4H3
Formel vikt 230.1154