Learn More
BD Difco™ Dehydrated Culture Media: Tryptic Soy Agar (Soybean-Casein Digest Agar)
Product Code. 11768822
Change view
Click to view available options
Quantity:
500 g
2 kg
10 kg
Packaging Type:
Bottle
Pail
Unit Size:
10kg
2kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Type Unit Size
11768822 500 g Bottle 500g
11748822 2 kg Pail 2kg
11758822 10 kg Pail 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11768822 Supplier BD Supplier No. 236950
 Restricted Item
Beräknad leverans: 09-12-2026
Use dehydrated media for isolation, detection and cultivation of a wide variety of microorganisms. BD Difco™ Dehydrated Culture Media: Tryptic Soy Agar (Soybean-Casein Digest Agar) is supplied in dehydrated form for the convenience of the laboratorian and bioprocessing specialist.
Applications:
  • Used with or without blood for cultivation and isolation of variety of organisms
  • Used for sterility testing
Certifications
AOAC, BAM, CCAM, COMPF, EP, EPA, ISO, JP, SMD, SMWW, USDA, USP
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance AOAC, BAM, CCAM, COMPF, EP, EPA, ISO, JP, SMD, SMWW, USDA, USP
Content And Storage 2°C to 30°C
Form Dehydrated
Media Type Tryptic Soy Agar
Product Type Tryptic Soy Agar
Formulation Pancreatic digest of Casein 15g/L, Papaic digest of Soybean 5g/L, Sodium Chloride 5g/L, Agar 15g/L
For Use With (Application) Isolation and cultivation of non-fastidious and fastidious microorganisms
Packaging Type Bottle
Quantity 500 g