Läs mer
Diflorasone Diacetate, TRC
Produktkod. 30180104
Change view
Kvantitet:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30180104 250 mg
30179747 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30180104 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD445500250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 33564-31-7
Molekylformel C26 H32 F2 O7
Molekylvikt (g/mol) 494.52
Synonym (6α,11β,16β)-17,21-Bis(acetyloxy)-6,9-difluoro-11-hydroxy-16-methylpregna-1,4-diene-3,20-dione, Dermaflor, Diacort, Diflorasone 17,21-diacetate, Diflorasone diacetate, Difulal, Florone, Maxiflor, Psorcon, Soriflor, U 34865
IUPAC-namn [2-[(6S,8S,9R,10S,11S,13S,14S,16S,17R)-17-acetyloxy-6,9-difluoro-11-hydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] acetate
LEDER C[C@H]1C[C@H]2[C@@H]3C[C@H](F)C4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(OC(=O)C)C(=O)COC(=O)C
Specifikationer
Kemiskt namn eller material Diflorasone Diacetate
Kvantitet 250 mg
InChI formel InChI=1S/C26H32F2O7/c1-13-8-17-18-10-20(27)19-9-16(31)6-7-23(19,4)25(18,28)21(32)11-24(17,5)26(13,35-15(3)30)22(33)12-34-14(2)29/h6-7,9,13,17-18,20-21,32H,8,10-12H2,1-5H3/t13-,17-,18-,20-,21-,23-,24-,25-,26-/m0/s1
Formel vikt 494.2116