Läs mer
Diflorasone Diacetate, TRC
Produktkod. 30373298
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30373298 10 mg
1 options
Produktkod. 30373298 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD44550010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Diflorasone Diacetate
CAS 33564-31-7
Molekylformel C26 H32 F2 O7
Kvantitet 10 mg
Synonym (6α,11β,16β)-17,21-Bis(acetyloxy)-6,9-difluoro-11-hydroxy-16-methylpregna-1,4-diene-3,20-dione, Dermaflor, Diacort, Diflorasone 17,21-diacetate, Diflorasone diacetate, Difulal, Florone, Maxiflor, Psorcon, Soriflor, U 34865
InChI formel InChI=1S/C26H32F2O7/c1-13-8-17-18-10-20(27)19-9-16(31)6-7-23(19,4)25(18,28)21(32)11-24(17,5)26(13,35-15(3)30)22(33)12-34-14(2)29/h6-7,9,13,17-18,20-21,32H,8,10-12H2,1-5H3/t13-,17-,18-,20-,21-,23-,24-,25-,26-/m0/s1
LEDER C[C@H]1C[C@H]2[C@@H]3C[C@H](F)C4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(OC(=O)C)C(=O)COC(=O)C
IUPAC-namn [2-[(6S,8S,9R,10S,11S,13S,14S,16S,17R)-17-acetyloxy-6,9-difluoro-11-hydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] acetate
Molekylvikt (g/mol) 494.52
Formel vikt 494.2116