missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Diflucortolone 21-Valerate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Diflucortolone Valerate |
| CAS | 59198-70-8 |
| Molekylformel | C27 H36 F2 O5 |
| Kvantitet | 50 mg |
| Synonym | Diflucortolone Valerate, (6α,11β,16α)-6,9-Difluoro-11-hydroxy-16-methyl-21-[(1-oxopentyl)oxy]pregna-1,4-diene-3,20-dione, Diflucortolone 21-valerate, Diflucortolone valerate, Neriforte, Nerisona, Nerisone, Nerisone Forte, Temetex, Texmeten, Travazol |
| InChI formel | InChI=1S/C27H36F2O5/c1-5-6-7-23(33)34-14-21(31)24-15(2)10-17-18-12-20(28)19-11-16(30)8-9-26(19,4)27(18,29)22(32)13-25(17,24)3/h8-9,11,15,17-18,20,22,24,32H,5-7,10,12-14H2,1-4H3/t15-,17+,18+,20+,22+,24-,25+,26+,27+/m1/s1 |
| LEDER | CCCCC(=O)OCC(=O)[C@H]1[C@H](C)C[C@H]2[C@@H]3C[C@H](F)C4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C |
| IUPAC-namn | [2-[(6S,8S,9R,10S,11S,13S,14S,16R,17S)-6,9-difluoro-11-hydroxy-10,13,16-trimethyl-3-oxo-7,8,11,12,14,15,16,17-octahydro-6H-cyclopenta[a]phenanthren-17-yl]-2-oxoethyl] pentanoate |
| Molekylvikt (g/mol) | 478.57 |
| Formel vikt | 478.2531 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?