Läs mer
Diflufenican, TRC
Product Code. 30370642
Change view
Click to view available options
Kvantitet:
1 g
Product Code. Kvantitet
30370642 1 g
1 options
Product Code. 30370642 Supplier Toronto Research Chemicals Supplier No. TRCD4456401G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Kemiskt namn eller material Diflufenican
CAS 83164-33-4
Molekylformel C19 H11 F5 N2 O2
Kvantitet 1 g
Synonym N-(2,4-Difluorophenyl)-2-[3-(trifluoromethyl)phenoxy]-3-pyridinecarboxamide, Brodal, Canyon, Diflufenican, Diflufenicanil, FR 1078, Hurricane (herbicide), Kwarc, Legato
InChI formel InChI=1S/C19H11F5N2O2/c20-12-6-7-16(15(21)10-12)26-17(27)14-5-2-8-25-18(14)28-13-4-1-3-11(9-13)19(22,23)24/h1-10H,(H,26,27)
LEDER Fc1ccc(NC(=O)c2cccnc2Oc3cccc(c3)C(F)(F)F)c(F)c1
IUPAC-namn N-(2,4-difluorophenyl)-2-[3-(trifluoromethyl)phenoxy]pyridine-3-carboxamide
Molekylvikt (g/mol) 394.29
Formel vikt 394.0741