Learn More
Bellingham Stanley™ Digital Refractometer RFM340-M
Product Code. 16339693
Change view
Click to view available options
100 to 240 V, 50/60 Hz
Product Code.
16339693 100 to 240 V, 50/60 Hz
1 options
Product Code. 16339693 Supplier Bellingham Stanley™ Supplier No. 1945
  • ONLINE EXCLUSIVE PRICE
    127890.00 SEK
    116011.45 SEK / Paket med 1
    Save 11878.55 SEK
    9% Av
View all promotions
Beräknad leverans: 29-12-2026
The Bellingham Stanley™ RFM340-M digital refractometer with three-decimal/0-100 Brix (1.58 RI) scales and Peltier control. Perfect for measurement and control in demanding food, beverage, chemical and industrial applications
  • Benchtop model for demanding food, beverage, chemical and industrial Brix or RI applications
  • Tactile and robust keypad with large 10 cm high resolution screen display
  • One-touch configuration of scale, temperature and data storage
  • User-defined methods or selected from the on-board library
  • Flat prism surface for easy cleaning
  • Wide beam scan for non-homogenous samples
TRUSTED_SUSTAINABILITY
Specifications
Type Benchtop Refractometer
Scale Range 0 to 100 Brix
Refractive Index 1.32 to 1.58
Accuracy ±0.00002 (1.32 to 1.38 RI), ±0.00004 (1.38 to 1.58 RI); ±0.010 (0 to 30°Brix), ±0.030 (30 to 100°Brix)
Resolution 0.000001 RI, 0.01/0.001 Brix
Measurement Time 4 sec.
Interface USB, Ethernet, RS232
Temperature Range (Metric) 0°C or 10°C below ambient whichever is greater to 70°C
Electrical Requirements 100/240 V 50/60 Hz