Läs mer
Digoxigenin Tyramide, TRC
Produktkod. 30369285
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30369285 10 mg
30373876 5 mg
30372607 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30369285 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD44651110MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 210231-99-5
Molekylformel C39H56N2O8
Molekylvikt (g/mol) 680.87
Synonym (3beta,5beta,12beta)-12,14-Dihydroxy-3-[2-[[6-[[2-(4-hydroxyphenyl)ethyl]amino]-6-oxohexyl]amino]-2-oxoethoxy]card-20(22)-enolide
IUPAC-namn 6-[[2-[[(3S,5R,8R,9S,10S,12R,13S,14S,17R)-12,14-dihydroxy-10,13-dimethyl-17-(5-oxo-2H-furan-3-yl)-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-3-yl]oxy]acetyl]amino]-N-[2-(4-hydroxyphenyl)ethyl]hexanamide
LEDER C[C@]12CC[C@@H](C[C@H]1CC[C@@H]3[C@@H]2C[C@@H](O)[C@]4(C)[C@H](CC[C@]34O)C5=CC(=O)OC5)OCC(=O)NCCCCCC(=O)NCCc6ccc(O)cc6
Specifikationer
Kemiskt namn eller material Digoxigenin Tyramide
Kvantitet 10 mg
InChI formel InChI=1S/C39H56N2O8/c1-37-16-13-29(48-24-35(45)40-18-5-3-4-6-34(44)41-19-15-25-7-10-28(42)11-8-25)21-27(37)9-12-31-32(37)22-33(43)38(2)30(14-17-39(31,38)47)26-20-36(46)49-23-26/h7-8,10-11,20,27,29-33,42-43,47H,3-6,9,12-19,21-24H2,1-2H3,(H,40,45)(H,41,44)/t27-,29+,30-,31-,32+,33-,37+,38+,39+/m1/s1
Formel vikt 680.4037