Läs mer
Dihexylphthalate, TRC
Produktkod. 30179621
Change view
Kvantitet:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30179621 500 mg
30179170 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30179621 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB185430500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 68515-50-4
Molekylformel C20H30O4
Molekylvikt (g/mol) 334.45
Synonym Phthalic acid bis-iso-hexyl ester, 1,2-Benzenedicarboxylic acid, 1,2-dihexyl ester (branched and linear), 1,2-Benzenedicarboxylic acid, dihexyl ester, branched and linear
IUPAC-namn 2-O-hexan-3-yl 1-O-(3-methylpentyl) benzene-1,2-dicarboxylate
LEDER CCCC(CC)OC(=O)c1ccccc1C(=O)OCCC(C)CC
Specifikationer
Kemiskt namn eller material Phthalic acid, bis-iso-hexyl ester
Analyt- eller komponentnamn Phthalic acid, bis-iso-hexyl ester
Anteckningar om renhetsgrad HPLC
Frakt skick Room Temperature
InChI formel InChI=1S/C20H30O4/c1-5-10-16(7-3)24-20(22)18-12-9-8-11-17(18)19(21)23-14-13-15(4)6-2/h8-9,11-12,15-16H,5-7,10,13-14H2,1-4H3
Lösningstyp Neat
Industry Type Pharmaceutical
Kvantitet 500 mg
Formel vikt 334.21
Procent renhet >95%