Läs mer
Dihydro Capsaicin, TRC
Produktkod. 30009155
Change view
Kvantitet:
10 mg
50 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30009155 250 mg
18031530 50 mg
18001840 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30009155 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD448750250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 19408-84-5
Molekylformel C18 H29 N O3
Molekylvikt (g/mol) 307.43
Synonym Dihydrocapsaicin, Nonanamide, 8-methyl-N-vanillyl- (7CI,8CI), 6,7-Dihydrocapsaicin, Capsaicin, dihydro-, Dihydrocapsaicin
IUPAC-namn N-[(4-hydroxy-3-methoxyphenyl)methyl]-8-methylnonanamide
LEDER COc1cc(CNC(=O)CCCCCCC(C)C)ccc1O
Specifikationer
Kemiskt namn eller material Dihydrocapsaicin
Anteckningar om renhetsgrad HPLC
Kvantitet 250 mg
InChI formel InChI=1S/C18H29NO3/c1-14(2)8-6-4-5-7-9-18(21)19-13-15-10-11-16(20)17(12-15)22-3/h10-12,14,20H,4-9,13H2,1-3H3,(H,19,21)
Formel vikt 307.2147
Procent renhet >95%