Läs mer
Dihydro Dutasteride, TRC
Produktkod. 30008337
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30008337 100 mg
18011030 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30008337 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD449870100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 164656-22-8
Molekylformel C27 H32 F6 N2 O2
Molekylvikt (g/mol) 530.55
Synonym 1H-Indeno[5,4-f]quinoline-7-carboxamide, N-[2,5-bis(trifluoromethyl)phenyl]hexadecahydro-4a,6a-dimethyl-2-oxo-, (4aR,4bS,6aS,7S,9aS,9bS,11aR)-, 4-Azaandrostane-17-carboxamide, N-[2,5-bis(trifluoromethyl)phenyl]-3-oxo-, (5α,17β)-, (5α,17β)-N-[(2,5-Bis(trifluoromethyl)phenyl]-3-oxo-4-aza-5-androstane-17-carboxamide, 1H-Indeno[5,4-f]quinoline-7-carboxamide, N-[2,5-bis(trifluoromethyl)phenyl]hexadecahydro-4a,6a-dimethyl-2-oxo-, [4aR-(4aα,4bβ,6aα,7α,9aβ,9bα,11aβ)]-, Dihydrodutasteride, N-[2,5-Bis(trifluoromethyl)phenyl]-3-oxo-4-aza-5α-androstane-17β-carboxamide
IUPAC-namn (1S,3aS,3bS,5aR,9aR,9bS,11aS)-N-[2,5-bis(trifluoromethyl)phenyl]-9a,11a-dimethyl-7-oxo-1,2,3,3a,3b,4,5,5a,6,8,9,9b,10,11-tetradecahydroindeno[5,4-f]quinoline-1-carboxamide
LEDER C[C@]12CCC(=O)N[C@@H]1CC[C@H]3[C@@H]4CC[C@H](C(=O)Nc5cc(ccc5C(F)(F)F)C(F)(F)F)[C@@]4(C)CC[C@H]23
Specifikationer
Kemiskt namn eller material Dihydro Dutasteride
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C27H32F6N2O2/c1-24-11-9-17-15(4-8-21-25(17,2)12-10-22(36)35-21)16(24)6-7-19(24)23(37)34-20-13-14(26(28,29)30)3-5-18(20)27(31,32)33/h3,5,13,15-17,19,21H,4,6-12H2,1-2H3,(H,34,37)(H,35,36)/t15-,16-,17-,19+,21+,24-,25+/m0/s1
Formel vikt 530.2368
Procent renhet >95%