Läs mer
Dihydro Ferulic Acid Methyl Ester, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Dihydro Ferulic Acid Methyl Ester |
| CAS | 56024-44-3 |
| Molekylformel | C11 H14 O4 |
| Kvantitet | 500 mg |
| Synonym | Benzenepropanoic acid, 4-hydroxy-3-methoxy-, methyl ester, Hydrocinnamic acid, 4-hydroxy-3-methoxy-, methyl ester (6CI), Methyl 3-(4-hydroxy-3-methoxyphenyl)propanoate, Methyl 3-[4-hydroxy-3-(methyloxy)phenyl]propanoate, Methyl dihydroferulate, Methyl hydroferulate, Dihydro ferulic acid methyl ester, 3-(4-Hydroxy-3-methoxyphenyl)propanoic acid methyl ester |
| InChI formel | InChI=1S/C11H14O4/c1-14-10-7-8(3-5-9(10)12)4-6-11(13)15-2/h3,5,7,12H,4,6H2,1-2H3 |
| LEDER | COC(=O)CCc1ccc(O)c(OC)c1 |
| IUPAC-namn | methyl 3-(4-hydroxy-3-methoxyphenyl)propanoate |
| Molekylvikt (g/mol) | 210.23 |
| Formel vikt | 210.0892 |